Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128180.1 | NP_001121652.1 | 7 - 8 | 1672 | 98 |
NM_001128181.1 | NP_001121653.1 | 8 - 9 | 1674 | 98 | |
NM_027712.3 | NP_081988.3 | 8 - 9 | 2659 | 98 | |
NM_177639.6 | NP_808307.2 | 9 - 10 | 2689 | 98 | |
GenBank mRNA | AK034182.1 | - | 3 - 4 | 548 | 98 |
AK035763.1 | - | 12 - 13 | 3240 | 98 | |
AK136926.1 | - | 8 - 9 | 1635 | 98 | |
AK147448.1 | - | 12 - 13 | 3476 | 98 | |
AK161941.1 | - | 7 - 8 | 1647 | 98 | |
AK220515.1 | - | 11 - 12 | 3539 | 98 | |
AY243846.2 | - | 8 - 9 | 2440 | 98 | |
BC062120.1 | - | 9 - 10 | 2690 | 98 | |
BC094369.1 | - | 7 - 8 | 1868 | 98 |