Assay Design



Gene Transcripts

Gene Symbol
Ganab
Entrez Gene ID
14376
Gene Name
alpha glucosidase 2 alpha neutral subunit
Gene Aliases
AU042638, G2an, GluII, mKIAA0088
Chromosome Location
Chr.19: 8898057 - 8916742 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293621.1 NP_001280550.1 15 - 16 1901 77
NM_008060.2 NP_032086.1 16 - 17 1967 77
GenBank mRNA AK017873.1 - 3 - 4 307 77
AK028655.1 - 15 - 16 1871 77
AK030722.1 - 15 - 16 1870 77
AK081915.1 - 15 - 16 1863 77
AK122201.1 - 10 - 11 2097 77
AK149686.1 - 16 - 17 1924 77
AK166150.1 - 15 - 16 1840 77
BC020386.1 - 8 - 9 1117 77
BC035423.1 - 6 - 7 717 77
BC094437.1 - 15 - 16 1851 77
BC117888.1 - 15 - 16 1844 77
BC117889.1 - 15 - 16 1847 77
BC128069.1 - 16 - 17 1911 77
U92793.1 - 16 - 17 1913 77

View Full Product Details