Assay Design



Gene Transcripts

Gene Symbol
Tut1
Entrez Gene ID
70044
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
2700038E08Rik, PAPD2, Rbm21, TUTase6
Chromosome Location
Chr.19: 8953850 - 8966210 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_197993.2 NP_932110.1 1 - 2 173 64
GenBank mRNA AK138418.1 - 1 - 2 93 64
BC023900.1 - 1 - 2 150 64
BC025499.1 - 1 - 2 197 64

View Full Product Details