Assay Design



Gene Transcripts

Gene Symbol
Ankrd13d
Entrez Gene ID
68423
Gene Name
ankyrin repeat domain 13 family, member D
Gene Aliases
0710001P18Rik, AI430801
Chromosome Location
Chr.19: 4270180 - 4283482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026720.2 NP_080996.2 11 - 12 1399 85
GenBank mRNA AK089186.1 - 11 - 12 1392 85
BC058982.1 - 11 - 12 1310 85

View Full Product Details