Assay Design



Gene Transcripts

Gene Symbol
Arhgap12
Entrez Gene ID
75415
Gene Name
Rho GTPase activating protein 12
Gene Aliases
2810011M08Rik
Chromosome Location
Chr.18: 6024448 - 6136098 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039692.1 NP_001034781.1 13 - 14 2216 121
NM_029277.2 NP_083553.2 12 - 13 2141 121
GenBank mRNA AK031648.1 - 13 - 14 2060 121
AK077063.1 - 13 - 14 2217 121
BC024633.1 - 8 - 9 628 121
BC037608.1 - 12 - 13 1956 121
BC141184.1 - 12 - 13 1950 121
BC145506.1 - 12 - 13 1949 121

View Full Product Details