Assay Design



Gene Transcripts

Gene Symbol
4930568A12Rik
Entrez Gene ID
75883
Gene Name
RIKEN cDNA 4930568A12 gene
Gene Aliases
-
Chromosome Location
Chr.1: 34477167 - 34499219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK016244.1 - 4 - 5 497 104


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI646763.1 - 1 - 2 Chr. 1 - 34477167 - 34480225 [+] on Build GRCm38 104
GenBankmRNA BX635597.1 - 1 - 2 Chr. 1 - 34477370 - 34480631 [+] on Build GRCm38 104
GenBankmRNA BY715855.1 - 495 Chr. - - [-] on Build 104


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream protein tyrosine phosphatase, non-receptor type 18 Ptpn18 FLP1, HSCF, PTP-HSCF, PTP-K1, Ptpk1 -3.6kb Sense Chr. - 34459746 - 34473784 on Build GRCm38
Downstream protease, serine 39 Prss39 Tesp1 18.2kb Sense Chr. - 34498430 - 34503062 on Build GRCm38

View Full Product Details