Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008428.3 | NP_001008428.1 | 4 - 5 | 461 | 65 |
GenBank mRNA | AK135785.1 | - | 4 - 5 | 442 | 65 |
AK136200.1 | - | 4 - 5 | 439 | 65 | |
BC085247.1 | - | 4 - 5 | 442 | 65 |