Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025692.3 | NP_079968.2 | 10 - 11 | 1191 | 97 |
GenBank mRNA | AK017787.1 | - | 10 - 11 | 1191 | 97 |
AK077825.1 | - | 10 - 11 | 1172 | 97 | |
AK151823.1 | - | 10 - 11 | 1148 | 97 | |
AK152029.1 | - | 10 - 11 | 1186 | 97 | |
AK168907.1 | - | 10 - 11 | 1185 | 97 | |
AK178397.1 | - | 3 - 4 | 282 | 97 | |
BC019764.1 | - | 10 - 11 | 1068 | 97 |