Assay Design



Gene Transcripts

Gene Symbol
Hnrnpul2
Entrez Gene ID
68693
Gene Name
heterogeneous nuclear ribonucleoprotein U-like 2
Gene Aliases
1110031M08Rik, AI465155, Hnrpul2
Chromosome Location
Chr.19: 8819401 - 8834142 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081196.1 NP_001074665.1 13 - 14 3038 120
GenBank mRNA DQ000354.1 - 13 - 14 2157 120

View Full Product Details