Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_146093.1 | NP_666205.1 | 7 - 8 | 594 | 110 |
GenBank mRNA | AK150442.1 | - | 6 - 7 | 597 | 110 |
AK159708.1 | - | 6 - 7 | 570 | 110 | |
AK185583.1 | - | 2 - 3 | 101 | 110 | |
BC006701.1 | - | 7 - 8 | 594 | 110 |