Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164634.1 | NP_001158106.1 | 2 - 3 | 555 | 67 |
NM_001164635.1 | NP_001158107.1 | 2 - 3 | 463 | 67 | |
NM_031194.5 | NP_112471.3 | 3 - 4 | 635 | 67 | |
GenBank mRNA | AB079895.1 | - | 1 - 2 | 292 | 67 |
AF078869.1 | - | 2 - 3 | 410 | 67 | |
AK031962.1 | - | 3 - 4 | 601 | 67 | |
AK044336.1 | - | 2 - 3 | 432 | 67 | |
AK157891.1 | - | 2 - 3 | 464 | 67 | |
BC014762.1 | - | 2 - 3 | 441 | 67 | |
BU615016.1 | - | 2 - 3 | 505 | 67 |