Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011778.2 | NP_035908.1 | 3 - 4 | 406 | 73 |
GenBank mRNA | AF143956.1 | - | 3 - 4 | 413 | 73 |
AK149639.1 | - | 3 - 4 | 416 | 73 | |
AK159329.1 | - | 1 - 2 | 100 | 73 | |
AK209750.1 | - | 2 - 3 | 275 | 73 | |
AK213023.1 | - | 2 - 3 | 275 | 73 | |
BC029326.1 | - | 3 - 4 | 365 | 73 | |
BC131944.1 | - | 3 - 4 | 353 | 73 | |
BC132617.1 | - | 3 - 4 | 353 | 73 |