Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194334.2 | NP_919315.2 | 5 - 6 | 1156 | 72 |
GenBank mRNA | AK089175.1 | - | 5 - 6 | 1122 | 72 |
AK129273.1 | - | 5 - 6 | 1142 | 72 | |
AK156788.1 | - | 5 - 6 | 1143 | 72 |