Assay Design



Gene Transcripts

Gene Symbol
Pcnxl3
Entrez Gene ID
104401
Gene Name
pecanex-like 3 (Drosophila)
Gene Aliases
mKIAA4073
Chromosome Location
Chr.19: 5664635 - 5688908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144868.3 NP_659117.2 31 - 32 5942 54
GenBank mRNA AF237953.1 - 29 - 30 3959 54
AK144114.1 - 7 - 8 1202 54
AK220471.1 - 9 - 10 1840 54
BC038056.1 - 26 - 27 4050 54
BC054751.1 - 31 - 32 5221 54
BC055289.1 - 32 - 33 5745 54
BC096538.1 - 31 - 32 5942 54

View Full Product Details