Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168488.1 | NP_001161960.1 | 4 - 5 | 893 | 70 |
NM_001168489.1 | NP_001161961.1 | 4 - 5 | 871 | 70 | |
NM_001168490.1 | NP_001161962.1 | 3 - 4 | 1207 | 70 | |
NM_008583.2 | NP_032609.1 | 3 - 4 | 1207 | 70 | |
GenBank mRNA | AB023401.1 | - | 3 - 4 | 834 | 70 |
AF016398.1 | - | 4 - 5 | 846 | 70 | |
AF072755.1 | - | 4 - 5 | 830 | 70 | |
AF109389.1 | - | 3 - 4 | 1206 | 70 | |
AF130368.1 | - | 3 - 4 | 1189 | 70 | |
AK133984.1 | - | 4 - 5 | 830 | 70 | |
AK154371.1 | - | 4 - 5 | 852 | 70 | |
AK164700.1 | - | 4 - 5 | 828 | 70 | |
AK170713.1 | - | 3 - 4 | 1187 | 70 | |
BC006610.1 | - | 4 - 5 | 826 | 70 | |
BC036287.1 | - | 3 - 4 | 1135 | 70 |