Assay Design
Gene Transcripts
- Gene Symbol
- Dnajc4
- Entrez Gene ID
-
57431
- Gene Name
- DnaJ (Hsp40) homolog, subfamily C, member 4
- Gene Aliases
-
2010301J22Rik, Hspf2, Mcg18
- Chromosome Location
-
Chr.19: 6987911 - 6992696 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK008507.1 |
- |
3 - 4 |
641 |
87 |
AK170008.1 |
- |
3 - 4 |
634 |
87 |
BC049281.1 |
- |
3 - 4 |
626 |
87 |
View Full Product Details