Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012021.2 | NP_036151.1 | 4 - 5 | 499 | 69 |
GenBank mRNA | AB041573.1 | - | 2 - 3 | 1401 | 69 |
AF110733.1 | - | 4 - 5 | 425 | 69 | |
AF124994.1 | - | 4 - 5 | 588 | 69 | |
AF197951.1 | - | 4 - 5 | 499 | 69 | |
AK002383.1 | - | 4 - 5 | 465 | 69 | |
AK003332.2 | - | 4 - 5 | 465 | 69 | |
AK009858.1 | - | 4 - 5 | 934 | 69 | |
AK136130.1 | - | 4 - 5 | 440 | 69 | |
AK152653.1 | - | 4 - 5 | 465 | 69 | |
BC008174.1 | - | 4 - 5 | 884 | 69 | |
BC014682.1 | - | 2 - 3 | 109 | 69 |