Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001194922.1 | NP_001181851.1 | 4 - 5 | 754 | 59 |
NM_001194923.1 | NP_001181852.1 | 4 - 5 | 691 | 59 | |
GenBank mRNA | AF349450.1 | - | 4 - 5 | 700 | 59 |
AF349453.1 | - | 4 - 5 | 692 | 59 |