Assay Design



Gene Transcripts

Gene Symbol
Faim
Entrez Gene ID
23873
Gene Name
Fas apoptotic inhibitory molecule
Gene Aliases
-
Chromosome Location
Chr.9: 98986373 - 99002019 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122851.1 NP_001116323.1 2 - 3 192 83
GenBank mRNA BU962258.1 - - 195 83

View Full Product Details