Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053159.3 | NP_444389.2 | 8 - 9 | 975 | 84 |
GenBank mRNA | AB049633.1 | - | 8 - 9 | 856 | 84 |
AK002509.1 | - | 8 - 9 | 945 | 84 | |
AK008592.1 | - | 8 - 9 | 1005 | 84 | |
AK054185.1 | - | 5 - 6 | 410 | 84 | |
AK077857.1 | - | 8 - 9 | 975 | 84 | |
AK145687.1 | - | 8 - 9 | 1005 | 84 | |
AK169272.1 | - | 8 - 9 | 976 | 84 | |
AK206417.1 | - | 1 - 2 | 49 | 84 | |
AK214851.1 | - | 1 - 2 | 49 | 84 | |
BC011128.1 | - | 8 - 9 | 944 | 84 | |
BC016586.1 | - | 8 - 9 | 848 | 84 | |
BC034877.1 | - | 8 - 9 | 941 | 84 | |
KC508086.1 | - | 8 - 9 | 952 | 84 |