Assay Design
Gene Transcripts
- Gene Symbol
- Prkar2a
- Entrez Gene ID
-
19087
- Gene Name
- protein kinase, cAMP dependent regulatory, type II alpha
- Gene Aliases
-
1110061A24Rik, AI317181, AI836829, RII(alpha)
- Chromosome Location
-
Chr.9: 108692143 - 108749512 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK013274.1 |
- |
1 - 2 |
565 |
70 |
View Full Product Details