Assay Design
Gene Transcripts
- Gene Symbol
- Cdhr4
- Entrez Gene ID
-
69398
- Gene Name
- cadherin-related family member 4
- Gene Aliases
-
Cdh29
- Chromosome Location
-
Chr.9: 107992492 - 107999683 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AI451251.1 |
- |
2 - 3 |
|
80 |
View Full Product Details