Assay Design



Gene Transcripts

Gene Symbol
Hyal3
Entrez Gene ID
109685
Gene Name
hyaluronoglucosaminidase 3
Gene Aliases
Hyl3
Chromosome Location
Chr.9: 107581296 - 107587359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178020.3 NP_821139.2 3 - 4 1042 90
GenBank mRNA AF074489.1 - 2 - 3 998 90
AY048681.1 - 3 - 4 1031 90
BC018457.1 - 3 - 4 1042 90

View Full Product Details