Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171004.1 | NP_001164475.1 | 17 - 18 | 3174 | 114 |
NM_001171005.1 | NP_001164476.1 | 16 - 17 | 2370 | 114 | |
NM_029239.3 | NP_083515.2 | 17 - 18 | 3171 | 114 | |
GenBank mRNA | AK030058.1 | - | 16 - 17 | 2366 | 114 |
BC023383.1 | - | 1 - 2 | 242 | 114 | |
BC027320.1 | - | 16 - 17 | 2573 | 114 | |
BC037012.1 | - | 16 - 17 | 2815 | 114 | |
BC043660.1 | - | 3 - 4 | 484 | 114 | |
BC089164.1 | - | 17 - 18 | 3073 | 114 |