Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242389.1 | NP_001229318.1 | 1 - 2 | 120 | 60 |
NM_001242390.1 | NP_001229319.1 | 1 - 2 | 120 | 60 | |
NM_001242391.1 | NP_001229320.1 | 1 - 2 | 120 | 60 | |
NM_134125.4 | NP_598886.1 | 1 - 2 | 120 | 60 | |
GenBank mRNA | AK088909.1 | - | 1 - 2 | 70 | 60 |
AK149902.1 | - | 1 - 2 | 92 | 60 | |
AY081142.1 | - | 1 - 2 | 71 | 60 | |
BC003249.1 | - | 1 - 2 | 122 | 60 |