Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161362.3 | NP_001154834.1 | 6 - 7 | 3150 | 86 |
NM_172144.3 | NP_742156.2 | 5 - 6 | 990 | 86 | |
GenBank mRNA | AK035077.1 | - | 5 - 6 | 770 | 86 |
AK076163.1 | - | 5 - 6 | 2202 | 86 | |
AK132175.1 | - | 5 - 6 | 991 | 86 | |
BC042841.1 | - | 5 - 6 | 2111 | 86 | |
BC042842.1 | - | 5 - 6 | 2584 | 86 | |
BC151170.1 | - | 5 - 6 | 2977 | 86 | |
BC151171.1 | - | 5 - 6 | 2972 | 86 |