Assay Design
Gene Transcripts
- Gene Symbol
- Camkmt
- Entrez Gene ID
-
73582
- Gene Name
- calmodulin-lysine N-methyltransferase
- Gene Aliases
-
1700106N22Rik, AI480743, AV099781, CLNMT, CaM KMT
- Chromosome Location
-
Chr.17: 85090592 - 85458589 on Build GRCm38
- Gene Symbol
- Gnao1
- Entrez Gene ID
-
14681
- Gene Name
- guanine nucleotide binding protein, alpha O
- Gene Aliases
-
AW050213, Galphao, Gnao, alphaO
- Chromosome Location
-
Chr.8: 93810561 - 93969388 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK147783.1 |
- |
- |
|
111 |
View Full Product Details