Assay Design



Gene Transcripts

Gene Symbol
Txnrd1
Entrez Gene ID
50493
Gene Name
thioredoxin reductase 1
Gene Aliases
TR, TR1, TrxR1
Chromosome Location
Chr.10: 82833951 - 82897724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042513.1 NP_001035978.1 2 - 3 204 58
NM_001042514.1 NP_001035979.1 1 - 2 292 58
GenBank mRNA AK011902.1 - 2 - 3 205 58
AK149625.1 - 2 - 3 205 58
AK151927.1 - 2 - 3 205 58
AK168356.1 - 2 - 3 205 58
AK172383.1 - 1 - 2 130 58
BC020274.1 - 1 - 2 59 58
BC037643.1 - 1 - 2 377 58
CJ136679.1 - 2 - 3 205 58
CX562215.1 - 1 - 2 292 58

View Full Product Details