Assay Design



Gene Transcripts

Gene Symbol
Dhx57
Entrez Gene ID
106794
Gene Name
DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57
Gene Aliases
AW494914
Chromosome Location
Chr.17: 80238304 - 80290476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163759.1 NP_001157231.1 14 - 15 3074 94
NM_198942.2 NP_945180.2 13 - 14 2915 94
GenBank mRNA AK053628.1 - 14 - 15 2742 94
BC026474.1 - 1 - 2 56 94
BC037692.1 - 2 - 3 151 94
BC062952.1 - 13 - 14 2915 94
BC065169.1 - 14 - 15 2741 94
BC066091.1 - 13 - 14 2577 94
BC079618.1 - 13 - 14 1854 94

View Full Product Details