Assay Design



Gene Transcripts

Gene Symbol
Dhx57
Entrez Gene ID
106794
Gene Name
DEAH (Asp-Glu-Ala-Asp/His) box polypeptide 57
Gene Aliases
AW494914
Chromosome Location
Chr.17: 80238304 - 80290476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163759.1 NP_001157231.1 20 - 21 4036 104
NM_198942.2 NP_945180.2 19 - 20 3877 104
GenBank mRNA AK035344.1 - 4 - 5 718 104
AK162192.1 - 2 - 3 280 104
BC026474.1 - 7 - 8 1018 104
BC037692.1 - 8 - 9 1113 104
BC062952.1 - 19 - 20 3877 104
BC065169.1 - 20 - 21 3703 104
BC066091.1 - 19 - 20 3539 104
BC079618.1 - 19 - 20 4157 104

View Full Product Details