Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163759.1 | NP_001157231.1 | 12 - 13 | 2857 | 136 |
NM_198942.2 | NP_945180.2 | 11 - 12 | 2698 | 136 | |
GenBank mRNA | AK053628.1 | - | 12 - 13 | 2525 | 136 |
BC062952.1 | - | 11 - 12 | 2698 | 136 | |
BC065169.1 | - | 12 - 13 | 2524 | 136 | |
BC066091.1 | - | 11 - 12 | 2360 | 136 | |
BC079618.1 | - | 11 - 12 | 1637 | 136 |