Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198168.3 | NP_937811.1 | 2 - 3 | 794 | 72 |
GenBank mRNA | AK034167.1 | - | 2 - 3 | 813 | 72 |
AK155872.1 | - | 2 - 3 | 786 | 72 | |
AK171041.1 | - | 2 - 3 | 765 | 72 | |
AK171178.1 | - | 2 - 3 | 795 | 72 | |
AK171430.1 | - | 2 - 3 | 803 | 72 | |
BC058977.1 | - | 2 - 3 | 585 | 72 |