Assay Design



Gene Transcripts

Gene Symbol
Mgat4c
Entrez Gene ID
67569
Gene Name
MGAT4 family, member C
Gene Aliases
9130411I17Rik, GntIVh
Chromosome Location
Chr.10: 101681487 - 102391851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162368.1 NP_001155840.1 2 - 3 253 153
GenBank mRNA BB611341.1 - 2 - 3 254 153

View Full Product Details