Assay Design



Gene Transcripts

Gene Symbol
Mgat4c
Entrez Gene ID
67569
Gene Name
MGAT4 family, member C
Gene Aliases
9130411I17Rik, GntIVh
Chromosome Location
Chr.10: 101681487 - 102391851 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162369.1 NP_001155841.1 1 - 2 80 140
GenBank mRNA AK032009.1 - 1 - 2 81 140

View Full Product Details