Assay Design



Gene Transcripts

Gene Symbol
Pus10
Entrez Gene ID
74467
Gene Name
pseudouridylate synthase 10
Gene Aliases
2810013G11Rik, 4933435A13Rik, AU014648, C77560, Ccdc139
Chromosome Location
Chr.11: 23665354 - 23732876 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033654.2 NP_001028826.1 10 - 11 941 135
NM_028304.2 NP_082580.1 10 - 11 1354 135
NM_028956.4 NP_083232.1 10 - 11 951 135
GenBank mRNA AK012737.1 - 10 - 11 1118 135
AK017063.1 - 10 - 11 928 135
AK028368.1 - 10 - 11 938 135
AK088562.1 - 10 - 11 942 135
AK145055.1 - 10 - 11 1300 135
BC025555.1 - 10 - 11 1172 135

View Full Product Details