Assay Design
Gene Transcripts
- Gene Symbol
- Rlim
- Entrez Gene ID
-
19820
- Gene Name
- ring finger protein, LIM domain interacting
- Gene Aliases
-
AL022832, AW743871, Ha1r, Rlim1, Rnf12
- Chromosome Location
-
Chr.X: 103957164 - 103981297 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_011276.3 |
NP_035406.3 |
2 - 3 |
284 |
135 |
GenBank mRNA |
AK029295.1 |
- |
2 - 3 |
278 |
135 |
View Full Product Details