Assay Design
Gene Transcripts
- Gene Symbol
- Pcnxl3
- Entrez Gene ID
-
104401
- Gene Name
- pecanex-like 3 (Drosophila)
- Gene Aliases
-
mKIAA4073
- Chromosome Location
-
Chr.19: 5664635 - 5688908 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF237953.1 |
- |
3 - 4 |
443 |
96 |
View Full Product Details