Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145516.2 | NP_663491.1 | 4 - 5 | 407 | 81 |
NM_175421.1 | NP_780630.1 | 4 - 5 | 407 | 81 | |
GenBank mRNA | AF189817.1 | - | 4 - 5 | 367 | 81 |
AK039771.1 | - | 4 - 5 | 375 | 81 | |
AK082118.1 | - | 4 - 5 | 360 | 81 | |
AK148653.1 | - | 4 - 5 | 390 | 81 | |
AK149868.1 | - | 4 - 5 | 367 | 81 | |
AK150074.1 | - | 4 - 5 | 367 | 81 | |
AK166091.1 | - | 4 - 5 | 375 | 81 | |
BC057994.1 | - | 4 - 5 | 375 | 81 | |
CK625615.1 | - | 4 - 5 | 407 | 81 |