Assay Design



Gene Transcripts

Gene Symbol
Thada
Entrez Gene ID
240174
Gene Name
thyroid adenoma associated
Gene Aliases
6530405K19, A930021O22, AU019899, BC052885, GITA
Chromosome Location
Chr.17: 84190055 - 84466261 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183021.3 NP_898842.2 21 - 22 3419 85
GenBank mRNA AK173258.1 - 6 - 7 929 85
EF222207.1 - 21 - 22 3327 85

View Full Product Details