Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285940.1 | NP_001272869.1 | 1 - 2 | 314 | 73 |
NM_001285946.1 | NP_001272875.1 | 1 - 2 | 314 | 73 | |
NM_001285949.1 | NP_001272878.1 | 1 - 2 | 314 | 73 | |
NM_199448.3 | NP_955519.1 | 1 - 2 | 314 | 73 | |
GenBank mRNA | AK033547.1 | - | 1 - 2 | 256 | 73 |
AK150241.1 | - | 1 - 2 | 280 | 73 | |
AK157221.1 | - | 1 - 2 | 281 | 73 | |
AK159545.1 | - | 1 - 2 | 267 | 73 | |
AK165595.1 | - | 1 - 2 | 264 | 73 | |
AY382580.1 | - | 1 - 2 | 265 | 73 | |
BC052366.1 | - | 1 - 2 | 293 | 73 | |
BC064130.1 | - | 1 - 2 | 220 | 73 | |
BC096619.1 | - | 1 - 2 | 259 | 73 |