Assay Design



Gene Transcripts

Gene Symbol
Fez2
Entrez Gene ID
225020
Gene Name
fasciculation and elongation protein zeta 2 (zygin II)
Gene Aliases
9030616F16, D17Ertd315e
Chromosome Location
Chr.17: 78369212 - 78418152 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285940.1 NP_001272869.1 6 - 7 1041 74
GenBank mRNA BC052366.1 - 6 - 7 1020 74

View Full Product Details