Assay Design
Gene Transcripts
- Gene Symbol
- Iqcf1
- Entrez Gene ID
-
74267
- Gene Name
- IQ motif containing F1
- Gene Aliases
-
1700055J15Rik
- Chromosome Location
-
Chr.9: 106499967 - 106502250 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_028843.1 |
NP_083119.1 |
1 - 2 |
79 |
75 |
GenBank mRNA |
AK006797.1 |
- |
1 - 2 |
55 |
75 |
View Full Product Details