Assay Design



Gene Transcripts

Gene Symbol
Gpr62
Entrez Gene ID
436090
Gene Name
G protein-coupled receptor 62
Gene Aliases
4933402E03Rik
Chromosome Location
Chr.9: 106463960 - 106466612 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159652.1 NP_001153124.1 1 - 1 237

View Full Product Details