Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175402.4 | NP_780611.3 | 1 - 1 | 2099 | 74 |
GenBank mRNA | AK075777.1 | - | 1 - 1 | 1042 | 74 |
BC052180.1 | - | 2 - 2 | 1281 | 74 | |
BC057609.1 | - | - | 38 | 74 | |
BC078452.1 | - | 2 - 2 | 1922 | 74 |