Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027643.1 | NP_081919.1 | 2 - 3 | 387 | 80 |
GenBank mRNA | AK033510.1 | - | 2 - 3 | 209 | 80 |
AK033896.1 | - | 2 - 3 | 218 | 80 | |
AK162625.1 | - | 2 - 3 | 221 | 80 | |
AK170548.1 | - | 2 - 3 | 213 | 80 | |
AK213481.1 | - | 1 - 2 | 114 | 80 | |
AK216197.1 | - | 1 - 2 | 115 | 80 | |
BC020020.1 | - | 2 - 3 | 238 | 80 |