Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019924.2 | NP_064308.2 | 12 - 13 | 1510 | 99 |
GenBank mRNA | AF074714.1 | - | 12 - 13 | 1482 | 99 |
AK004412.1 | - | 2 - 3 | 116 | 99 | |
AK089467.1 | - | 12 - 13 | 1502 | 99 | |
AK154674.1 | - | 12 - 13 | 1509 | 99 | |
BC012964.1 | - | 12 - 13 | 1520 | 99 |