Assay Design



Gene Transcripts

Gene Symbol
Ptprs
Entrez Gene ID
19280
Gene Name
protein tyrosine phosphatase, receptor type, S
Gene Aliases
AL022616, PTP, PTP-NU3, PTPNU-3, PTPsigma, Ptpt9, R-PTP-S, RPTPsigma
Chromosome Location
Chr.17: 56412426 - 56476480 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252453.1 NP_001239382.1 19 - 20 3210 94
NM_001252455.1 NP_001239384.1 18 - 19 3198 94
NM_001252456.1 NP_001239385.1 19 - 20 3187 94
NM_011218.2 NP_035348.2 23 - 24 4428 94
GenBank mRNA AK144123.1 - 11 - 12 1789 94
AK159320.1 - 6 - 7 491 94
AK169714.1 - 2 - 3 184 94
BC052462.1 - 19 - 20 3136 94
BC054077.1 - 19 - 20 3104 94
BC083188.1 - 18 - 19 3157 94
D28530.1 - 23 - 24 4317 94
X82288.1 - 22 - 23 4237 94

View Full Product Details