Assay Design
Gene Transcripts
- Gene Symbol
- Carns1
- Entrez Gene ID
-
107239
- Gene Name
- carnosine synthase 1
- Gene Aliases
-
AI790298, Atpgd1, mKIAA1394
- Chromosome Location
-
Chr.19: 4164324 - 4175479 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134148.2 |
NP_598909.2 |
5 - 6 |
952 |
97 |
View Full Product Details