Assay Design



Gene Transcripts

Gene Symbol
Acsbg2
Entrez Gene ID
328845
Gene Name
acyl-CoA synthetase bubblegum family member 2
Gene Aliases
Bgr
Chromosome Location
Chr.17: 56843103 - 56874504 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039114.1 NP_001034203.1 9 - 10 1385 82
GenBank mRNA DQ250679.1 - 9 - 10 1385 82

View Full Product Details