Assay Design



Gene Transcripts

Gene Symbol
Slc25a23
Entrez Gene ID
66972
Gene Name
solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 23
Gene Aliases
2310067G05Rik, SCaMC-3
Chromosome Location
Chr.17: 57043711 - 57059880 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025877.4 NP_080153.2 1 - 2 346 108
GenBank mRNA AK158821.1 - 1 - 2 129 108
AK206310.1 - 1 - 2 75 108
BC072660.1 - 1 - 2 342 108

View Full Product Details